J&W DB-Sulfur SCD Columns, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
G3903-63001 G3903-63002 G3903-63003 G3903-63004
CAAGG390363001EA 2312.6 CAD
CAAGG390363001 CAAGG390363002 CAAGG390363003 CAAGG390363004
J&W DB-Sulfur SCD Columns, Agilent Technologies
Chromatography Columns
Engineered for sulfur chemiluminescence detection (SCD) to provide low-bleed performance and reduced ceramic tube fouling.
  • Improved data reliability and significantly reduced bleed
  • Optimal SCD stability and lower bleed
  • Reduces downtime, maintenance, and operational cost
  • Available in custom configurations
  • Performance Summary shipped with each column
Agilent J&W DB-Sulfur SCD columns are optimized for use with sulfur chemiluminescence detection (SCD) to provide low bleed performance and reduced SCD ceramic tube fouling. For all GC/SCD methods thatMore Product Information