J&W DB-Petro Columns, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
122-10A6 128-1056
CA122-10A6EA 2265.66 CAD
CA122-10A6 CAAG128-1056
J&W DB-Petro Columns, Agilent Technologies
Chromatography Columns
Configured for the analysis of petroleum process products PONA, PIANO, and PNA.
  • 100% Dimethylpolysiloxane
  • Configured for the analysis of petroleum process products
  • PONA, PIANO, PNA
  • High resolution
  • Bonded, crosslinked, and solvent rinsable
  • Tested with the tightest industry QC specifications for column bleed, sensitivity, and efficiency
  • Performance Summary shipped with each column
Agilent J&W DB-Petro is a 100% dimethylpolysiloxane phase, precision engineered for the analysis of PONA, PIANO, and PNA from petroleum processes. These high-resolution columns are bonded, crosslinked, and solvent rinsable, and have been extensively QC tested to the strictest industry standards. DB-Petro also features an operating temperature range of –60 to 325/350 °C.