J&W DB-608 Columns, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
125-1730 125-6837 122-6832 123-1730
CA125-1730EA 1268.35 CAD
CA125-1730 CA125-6837 CAAG122-6832 CAAG123-1730
J&W DB-608 Columns, Agilent Technologies
Chromatography Columns
Specifically designed for the analysis of chlorinated pesticides and PCBs, suitable for use in EPA methods 608, 508, and 8080.
  • Specifically designed for the analysis of chlorinated pesticides and PCBs
  • US EPA Methods 608, 508, and 8080
  • Excellent inertness and recovery without pesticide breakdown
  • Bonded, crosslinked, and solvent rinsable
  • Tested with the tightest industry QC specifications for column bleed, sensitivity, and efficiency
  • Performance Summary shipped with each column
Agilent J&W DB-608 columns are precision-engineered to deliver industry-leading performance in the analysis of chlorinated pesticides and PCBs. Widely used for US EPA methods 608, 508, and 8080, they deliver excellent inertness and recovery, without resulting in pesticide breakdown. This column is bonded, crosslinked, and solvent rinsable, with operating temperature limits of 40 to 280/300 °C.