J&W DB-225 Columns, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
122-2212 122-2232 123-2232 122-2231 122-2232-INT 123-2232-INT 125-2212 125-2232 125-2237
CA122-2212EA 671.39 CAD
CA122-2212 CA122-2232 CA123-2232 CAAG122-2231 CAAG1222232INT CAAG1232232INT CAAG125-2212 CAAG125-2232 CAAG125-2237
J&W DB-225 Columns, Agilent Technologies
Chromatography Columns
Ideal for separations of cis- and trans-fatty acid methyl esters in food testing applications.
  • (50%-Cyanopropylphenyl)-dimethylpolysiloxane
  • Mid/high polarity
  • Excellent for separations of cis- and trans-fatty acid methyl esters (FAMEs)
  • Bonded, crosslinked, and solvent rinsable
  • Exact replacement of HP-225
  • Close equivalent to USP Phase G7
  • Tested with the tightest industry QC specifications for column bleed, sensitivity, and efficiency
  • Performance Summary shipped with each column
  • Similar phases: SP-2330, Rtx-225, BP-225, OV-225, 007-225, AT-225