J&W CP-SimDist Columns, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
CP7512 CP7569 CP7532 CP7542 CP7562 CP7570 CP7571 CP7582 CP7592
CACP7512EA 1534.95 CAD
CACP7512 CACP7569 CAAGCP7532 CAAGCP7542 CAAGCP7562 CAAGCP7570 CAAGCP7571 CAAGCP7582 CAAGCP7592
J&W CP-SimDist Columns, Agilent Technologies
Chromatography Columns
CP-SimDist UltiMetal performs extended analysis to C120, and is guaranteed to a Tmax of 450 °C, due to the virtually unbreakable UltiMetal deactivated stainless steel tubing.
  • Designed for ASTM D2887 and the extended D2887 method compliance
  • Low bleed
  • Extended analysis to C120 with maximum temperature of 450 °C
  • UltiMetal deactivated stainless steel tubing for excellent durability (same id as 0.53 mm id fused silica)
  • Excellent retention time repeatability and column lifetime due to special deactivation of UltiMetal surface
  • Tested with the tightest industry QC
More Product Information