ZORBAX for NP/SFC, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
820675-111 820950-901 820950-908 820950-919 821125-919 880952-201 880952-701 959759-901 959961-901 820668-901 820668-931 821725-938 827975-301 827975-901 828700-901 828975-901 860700-708 880952-205 880952-208 880953-901 883952-701 959743-901 959758-901 959793-901
CA820675-111PK 618.49 CAD
CA820675-111 CA820950-901 CA820950-908 CA820950-919 CA821125-919 CA880952-201 CA880952-701 CAAG959759-901 CAAG959961-901 CAAG820668-901 CAAG820668-931 CAAG821725-938 CAAG827975-301 CAAG827975-901 CAAG828700-901 CAAG828975-901 CAAG860700-708 CAAG880952-205 CAAG880952-208 CAAG880953-901 CAAG883952-701 CAAG959743-901 CAAG959758-901 CAAG959793-901
ZORBAX for NP/SFC, Agilent Technologies
Chromatography Columns
Fully porous columns based on ZORBAX silica for normal phase LC and SFC separations.
  • Traditional fully porous ZORBAX silica particles available with different stationary phases and particle sizes
  • Master challenging normal phase applications with orthogonal chemistries such as ZORBAX Rx-Sil, CN, and NH2 phases
  • Discover the robust, general-purpose ZORBAX HILIC Plus and Rx-Sil columns for SFC, available in 1.8 µm and 5 µm particle sizes
  • Use Agilent Application Finder to find application notes of ZORBAX columns used in NP-LC, SFC, and SFC/MS
Agilent ZORBAX fully porous particle columns for normal phase HPLC and supercritical fluid chromatography (SFC) are designed for the retention and separation of lipophilic compounds. The unique ZORBAX chemistries deliver outstanding efficiency in both achiral SFC and normal phase chromatography due to the similar retention mechanisms of both LC modes on the polar stationary phases used. The recommended ZORBAX phases are Rx-Sil, amino (NH2), cyano (CN), Eclipse XDB-CN, and HILIC Plus (HILIC phases in SFC only).