J&W Ultra Inert Intuvo Columns, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
122-0334UI-INT 123-0334UI-INT 122-9732-INT 123-9334UI-INT 123-9434UI-INT G3909-63004
CAAG1220334UNTEA 1692.93 CAD
CAAG1220334UNT CAAG1230334UNT CAAG1229732INT CAAG1239334UNT CAAG1239434UNT CAAGG390963004
J&W Ultra Inert Intuvo Columns, Agilent Technologies
Chromatography Columns
Provide excellent inertness, low-bleed performance, and detection limits with the Intuvo 9000 GC system.
  • Wide selection of stationary phase polarities are available (non, low, mid, and high polarity)
  • Strict quality control specifications and testing ensure long-term column-to-column performance and accurate separations
  • Optimized GC flow path with Agilent Ultra Inert Liners, low-bleed septa, certified vials, and gas clean filters
  • For use with the Agilent Intuvo 9000 GC System
As part of the Agilent J&W Ultra Inert GC column family, Ultra InertMore Product Information