J&W Intuvo Premium Polysiloxane Columns, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
122-1111-INT 122-5711-INT 122-5731-INT 123-1111-INT 123-57J1-INT CP7483-INT CP7596-INT G3909-63005 G3909-63006
CAAG1221111INTEA 1050.11 CAD
CAAG1221111INT CAAG1225711INT CAAG1225731INT CAAG1231111INT CAAG12357J1INT CAAGCP7483-INT CAAGCP7596-INT CAAGG390963005 CAAGG390963006
J&W Intuvo Premium Polysiloxane Columns, Agilent Technologies
Chromatography Columns
The most common stationary phases – stable, robust, and versatile; for use with the Intuvo 9000 GC System.
  • Wide selection of stationary phase polarities (low, mid, and high polarity)
  • Strict quality control specifications and testing ensure long-term column-to-column performance and accurate separations
  • Optimized GC flow path with Agilent Ultra Inert Liners, low-bleed septa, certified vials, and gas clean filters
  • For use with the Agilent Intuvo 9000 System
Agilent J&W Premium Polysiloxane Intuvo GC columns (compatible only with the Intuvo 9000 GC System) are most common stationary phases. Polysiloxanes are stable, robust, and versatile, and are available in the greatest variety. Standard polysiloxanes are characterized by the repeating siloxane backbone. Each silicon atom contains two functional groups; the type and percent level of substitution of the groups distinguish each stationary phase and its properties.