Diamond® APEX™ Pipettors, Multi-Channel, Globe Scientific
Supplier: Globe Scientific
3354-10 3354-20 3354-50 3354-100 3354-200 3354-300 3355-10 3355-20 3355-50 3355-100 3355-200 3355-300
76509-230EA 833.72 CAD
76509-230 76509-232 76509-234 76509-236 76509-238 76509-240 76509-242 76509-244 76509-246 76509-248 76509-250 76509-252
Diamond® APEX™ Pipettors, Multi-Channel, Globe Scientific
Pipettes Micropipettes
The Diamond® APEX™ 8-channel and 12-channel pipettors include the myriad of features found in the single channel models and more.
  • Each channel is calibrated individually
  • Removable, fully autoclavable and UV resistant manifold
  • Three-year warranty
  • Magnet assisted piston for extremely high accuracy and consistency
  • Ergonomic design and light weight
  • Precise volume adjustment
  • Locking mechanism to prevent accidental volume changes
Simple operation and light weight aid in reducing fatigue and a 360° rotating manifold allows for positioning vertically or horizontally while aspirating or dispensing. As with the single channel models, every multi-channel Diamond® APEX™ pipettor is calibrated in an ISO 17025 accredited facility and come standard with a three-year warranty.