J&W Ultra Inert DB-WAX Columns, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
122-7032UI 122-7033UI 123-7033UI 122-7062UI 122-7032UI-INT 123-7032UI 125-7032UI 122-7012UI 122-7063UI 122-7062UI-INT 123-7012UI 123-7062UI 123-7063UI 123-7032UI-INT 123-7033UI-INT 125-7012UI 125-7031UI 125-7037UI 128-7022UI
CA122-7032UIEA 1263.79 CAD
CA122-7032UI CA122-7033UI CA123-7033UI CAAG122-7062UI CAAG1227032UNT CAAG123-7032UI CAAG125-7032UI CAAG122-7012UI CAAG122-7063UI CAAG1227062UNT CAAG123-7012UI CAAG123-7062UI CAAG123-7063UI CAAG1237032UNT CAAG1237033UNT CAAG125-7012UI CAAG125-7031UI CAAG125-7037UI CAAG128-7022UI
J&W Ultra Inert DB-WAX Columns, Agilent Technologies
Chromatography Columns
Deliver the highest levels of PEG stationary phase inertness performance for a range of challenging active analytes.
  • Improved peakshape for challenging polar compounds with DB-WAX Ultra Inert
  • Reliable column-to-column inertness performance
  • Bonded polyethylene glycol (PEG) selectivity equivalent to DB-WAX
  • No method revalidation needed
  • High polarity
  • Equivalent to USP G16
  • Excellent performance for acidic compounds, including free fatty acids – no need for FFAP type column
  • Bonded and crosslinked phase is solvent rinseable and tolerates aqueous injections