DB-UI 8270D Columns, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
122-9732 122-9736
CA122-9732EA 1348.91 CAD
CA122-9732 CA122-9736
DB-UI 8270D Columns, Agilent Technologies
Chromatography Columns
Delivers excellent peak shapes for ultra-trace-level analysis of active semi-volatile organic compounds targeted by EPA 8270D.
  • Specifically engineered for improved trace-level analysis of active semi volatile organic compounds
  • Industry-leading Ultra Inert deactivation and manufacturing
  • Unmatched testing protocol provides ultimate confidence in your results
  • ‘MS-Grade’ low-bleed stationary phase ensures the lowest interference from the column at elevated temperatures
  • Tested with the tightest industry QC specifications for column bleed, sensitivity, and efficiency
  • Performance Summary shipped with each column
Agilent J&W DB-UI8270D delivers excellent peak shape for trace-level analysis of active semi volatile organic compounds targeted by EPA method 8270D. Leading-edge manufacturing processes, combined with optimized chemistries and design advancements, improve both inertness and column-to-column reproducibility with no trade-offs in bleed or stationary phase selectivity.
All DB-UI8270D Ultra Inert columns are individually tested with the most highly probative semi volatile compounds, including propanoic acid, pyridine, 1,2 butanediol, 1-chloro-2-fluorobenzene, and m/p xylene, at low temperature (45 °C), to ensure excellent peak shape performance at low levels (5 to 10 ng on-column).