J&W DB-HeavyWax Columns, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
121-7122 122-7132 122-7133 127-7112 122-7112 122-7127 122-7157 122-7162 122-7163 122-7132-INT 122-7162-INT 123-7112 123-7132 123-7133 123-7162 123-7163 123-7132-INT 123-7133-INT 123-7163-INT
CAAG121-7122EA 1076.52 CAD
CAAG121-7122 CAAG122-7132 CAAG122-7133 CAAG127-7112 CAAG122-7112 CAAG122-7127 CAAG122-7157 CAAG122-7162 CAAG122-7163 CAAG1227132INT CAAG1227162INT CAAG123-7112 CAAG123-7132 CAAG123-7133 CAAG123-7162 CAAG123-7163 CAAG1237132INT CAAG1237133INT CAAG1237163INT
J&W DB-HeavyWax Columns, Agilent Technologies
Chromatography Columns
A high-temperature WAX column that can operate at temperatures of 280 °C isothermal and 290 °C programmed.
  • Polyethylene glycol selectivity
  • Maximum operating temperatures of 280 °C isothermal and 290 °C programmed
  • Fast analysis, with shorter run times due to extended temperature limit
  • Retention time stability and increased column lifetime, even when used at maximum operating temperature
  • Ability to bake out the column at the end of the analysis to reduce sample matrix effect and limit carryover and the risk of ghost peaks
  • Extended list of analytes to higher
More Product Information