CrossLab DB-CLP 1 and DB-CLP 2 Columns, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
123-8232 123-8336
CA123-8232EA 1325.7 CAD
CA123-8232 CA123-8336
CrossLab DB-CLP 1 and DB-CLP 2 Columns, Agilent Technologies
Chromatography Columns
Agilent J&W DB-CLP1 and DB-CLP2 is a flexible universal column pair for complete resolution and confirmation of 22 Contract Lab Program (CLP) pesticides.
  • Universal column pair designed for pesticides analyses
  • EPA Methods: CLP (Contract Lab Program) pesticides, 504.1, 505, 508.1, 551, 552.3, 8081B, 8082A, 8154A
  • Ideal for dual-column, dual-ECD GC analyses
  • Column pairs can be easily connected with an Agilent Ultra Inert, universal press fit Y–splitter (5190-6980), or an UltiMetal Plus deactivated CFT unpurged splitter (G3184-60065)
  • Mid polarity stabilized phases provide fast and low-bleed reliable analyses
  • Special testing includes pesticides for proof of performance and column-to-column reproducibility for DB-CLP1 primary, DB-CLP2 confirmation
  • Tested with the tightest industry QC specifications for column bleed, sensitivity, and efficiency
  • Performance Summary shipped with each column
Agilent J&W DB-CLP1 and DB-CLP2 are the most flexible and universal column pair for nine EPA dual-ECD pesticide methods as well as the analysis of Contract Lab Program (CLP) pesticides. Together, these fast, reliable columns deliver excellent resolving power with exceptionally low bleed while eliminating the need for time-consuming column switching.
DB-CLP1 and DB-CLP2 GC columns are subjected to the industry’s most stringent testing procedures, and a performance summary sheet is supplied with every column.