J&W DB-624 Columns, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
122-1334 122-1334E 122-1364 122-1364E 123-1334E 123-1364 123-1364E 125-1334E 125-1364 125-1374 125-1374E 128-1324 123-1334 125-1334 121-1324E 123-1334-KEY 125-1364E 125-1334-KEY 122-1334-KEY 122-1364-KEY 123-1364-KEY 124-1334 124-1374 125-1314 125-13A4 125-13B4 125-1364-KEY 128-1324E
CA122-1334EA 1245.47 CAD
CA122-1334 CA122-1334E CA122-1364 CA122-1364E CA123-1334E CA123-1364 CA123-1364E CA125-1334E CA125-1364 CA125-1374 CA125-1374E CA128-1324 CA76514-550 CA76514-552 CAAG121-1324E CAAG1231334KEY CAAG125-1364E CAAG1251334KEY CAAG1221334KEY CAAG1221364KEY CAAG1231364KEY CAAG124-1334 CAAG124-1374 CAAG125-1314 CAAG125-13A4 CAAG125-13B4 CAAG1251364KEY CAAG128-1324E
J&W DB-624 Columns, Agilent Technologies
Chromatography Columns
Recommended for EPA methods 502.2 and 8021, as well as for fast GC/MS analysis of volatiles.
  • Specifically designed for the analysis of volatile priority pollutants and residual solvents
  • Bonded, crosslinked, and solvent rinsable
  • Equivalent to USP Phase G43
  • Exact replacement of HP-624
  • Available with DuraGuard (GC column with a built-in guard column)
  • No cryogenics needed for U.S. EPA Method 502.2
  • Excellent for U.S. EPA Methods 501.3, 502.2, 503.1, 524.2, 601, 602, 8010, 8015, 8020, 8240, 8260 and USP 467
  • Excellent inertness for active compounds
  • Tested with
More Product Information