DB-5ht Columns, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
122-5711 122-5731 123-5711 123-5731 122-5732 122-5711E 123-5701
CA122-5711EA 823.07 CAD
CA122-5711 CA122-5731 CA123-5711 CA123-5731 CAAG122-5732 CAAG122-5711E CAAG123-5701
DB-5ht Columns, Agilent Technologies
Chromatography Columns
A nonpolar column specially processed for an extended temperature limit of 400 °C.
  • (5%-Phenyl)-methylpolysiloxane
  • Nonpolar
  • Specially processed for extended temperature limit of 400 °C
  • High temperature, polyimide-coated, fused silica tubing
  • Excellent peak shape and faster elution times for high boilers
  • Bonded, crosslinked, and solvent rinsible
  • Tested with the tightest industry QC specifications for column bleed, sensitivity, and efficiency
  • Performance Summary shipped with each column
  • Similar phases: HT5, Stx-5ht, ZB-5ht
Agilent J&W DB-5ht nonpolarMore Product Information