J&W Ultra Inert DB-35ms Columns, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
122-3812UI 122-3832UI 121-3822UI 122-3812UI-INT 123-3832UI 122-3832UI-INT 123-3832UI-INT
CA122-3812UIEA 819.43 CAD
CA122-3812UI CA122-3832UI CAAG121-3822UI CAAG1223812UNT CAAG123-3832UI CAAG1223832UNT CAAG1233832UNT
J&W Ultra Inert DB-35ms Columns, Agilent Technologies
Chromatography Columns
Improved inertness and exceptionally low bleed, with great peak shapes for challenging active analytes such as pesticides and drugs of abuse.
  • Individually tested with the Ultra Inert DB-35ms test probe mixture
  • Consistent column inertness performance
  • Exceptionally low column bleed
  • Great peak shapes for challenging active analytes, with excellent signal-to-noise ratios
  • Minimum compound adsorption or degradation
  • Supports 0.18 mm int.Ø column configuration for higher sample throughput
Agilent J&W DB-35ms Ultra Inert is a mid-polarity column forMore Product Information