J&W DB-1ms Ultra Inert Columns, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
122-0132UI 122-0112UI 122-0162UI 122-0132UI-INT 123-0112UI 123-0132UI 123-0132UI-INT
CA122-0132UIEA 1349.8 CAD
CA122-0132UI CAAG122-0112UI CAAG122-0162UI CAAG1220132UNT CAAG123-0112UI CAAG123-0132UI CAAG1230132UNT
J&W DB-1ms Ultra Inert Columns, Agilent Technologies
Chromatography Columns
Provides consistent column inertness performance to ensure sensitivity and integrity in trace analysis.
  • Individually tested with Ultra Inert 1ms test probe mixture
  • Consistent column inertness performance and exceptionally low bleed
  • High-quality peak shapes for challenging active analytes
  • Excellent signal-to-noise ratios
  • Minimum compound adsorption or degradation
  • Supports 0.18 mm int.Ø column configuration for higher sample throughput
  • Performance Summary shipped with each column
Agilent J&W DB-1ms Ultra Inert delivers consistent column inertness performance to ensure analytical sensitivity and integrity. With excellent peak shapes, exceptionally low bleed, and high signal-to-noise ratios, this column performs effectively with even the most challenging active analytes. Each column is individually tested with Ultra Inert 1ms test probe mixture.
Tested against the strictest industry QC specifications for column bleed, sensitivity and efficiency.