J&W DB-1ht Columns, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
122-1111 122-1131 123-1111 123-1131 123-1102LTM 123-1131E 123-1111LTM 125-1131 G3903-61006
CA122-1111EA 823.07 CAD
CA122-1111 CA122-1131 CA123-1111 CA123-1131 CAAG1231102LTM CAAG123-1131E CAAG1231111LTM CAAG125-1131 CAAGG390361006
J&W DB-1ht Columns, Agilent Technologies
Chromatography Columns
100% PDMS specially processed for temperature limits of 400 °C, with excellent peak shape and faster elution times for high boilers.
  • 100% Dimethylpolysiloxane
  • Nonpolar
  • Specially processed for extended temperature limit of 400 °C
  • High temperature, polyimide-coated, fused silica tubing
  • Excellent peak shape and faster elution times for high boilers
  • Bonded, crosslinked, and solvent rinsable
  • Tested with the tightest industry QC specifications for column bleed, sensitivity, and efficiency
  • Performance Summary shipped with each column
Agilent J&W DB-1ht is a nonpolar, high-temperature 100% dimethlypolysiloxane column manufactured to the highest levels of quality and stability to deliver excellent performance in a range of GC/MS applications.It is specially processed for operation up to 400 °C, with a specially engineered high-temperature polyimide-coated fused silica tubing.
This bonded, crosslinked, solvent-rinsable column delivers excellent peak shape and faster elution times for high-boiling compounds.