J&W 8890 GC Columns with Smart Key, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
125-7032-KEY 19091J-413-KEY 122-5032-KEY 122-5532-KEY 122-5532UI-KEY 122-7032-KEY 19091S-433-KEY 19091S-431UI-KEY CP8944-KEY
CAAG1257032KEYEA 1440.12 CAD
CAAG1257032KEY CAAG19091J413K CAAG1225032KEY CAAG1225532KEY CAAG1225532UIK CAAG1227032KEY CAAG19091S433K CAAG19091S43UK CAAGCP8944-KEY
J&W 8890 GC Columns with Smart Key, Agilent Technologies
Chromatography Columns
Various GC columns with a Smart Key included, made specifically to fit the 8890 GC.
  • Smart Key allows self-configuration, usage monitoring, and help automating method development when using 8890 GC
  • Wide selection of available stationary phases (low, mid, and high polarity, application-specific stationary phases)
  • Strict quality control specifications and testing ensure long-term column-to-column performance and accurate separations
  • Optimized GC flow path with Agilent Ultra Inert Liners, low-bleed septa, certified vials, and gas clean filters
  • For use with the
More Product Information