VWR® Eco Premium ULT Freezers
Supplier: VWR International
9020-0450 9020-0468 9020-0467 9020-0462 9020-0469
76514-174EA 19624.04 CAD
76514-174 76514-176 76533-504 76533-506 76683-448 76683-450
VWR® Eco Premium ULT Freezers
Freezers Ultra Low Temperature Freezers
The VWR® ULT Freezers Eco Premium are designed for safe and reliable storage of sensitive samples. They feature ergonomic control and low energy consumption.
  • Maximum 2'' box capacity: 264/352/528
  • Energy star certified
  • Temperatures between −86 and −50 °C
  • Eco Premium 352 and 528: Long warm-up time (warm-up time from −86 and −50 °C: 360 min)
  • Eco Premium 264: Long warm-up time (warm-up time from −86 and −50 °C: 280 min)
  • Highly efficient VIP insulation and long lasting rust proof sample chamber
  • Green refrigerants
  • USB and Ethernet interface for data transfer
The VWR® ULT Freezers Eco Premium have a stainless steel inner chamber, flexible rack configuration and are further characterized by low energy consumption and electricity costs. They have individually adaptable chamber compartments and samples remain frozen for >30 hours during power outage or system failure. The climate-friendly refrigerants used help to protect the environment.
Warranty:
-VWR® Five-year limited parts and labor warranty
-VWR® Five-year warranty on compressor parts
Certifications: UL, cUL, CE marked, UKCA.
Ordering information: A variety of accessories are available, these must be ordered separately.