CrossLab Peristaltic Pump Tubing for MP-AES, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
3710067900 G8003-67003
CA3710067900PK 133.81 CAD
CA3710067900 CAAGG800367003
CrossLab Peristaltic Pump Tubing for MP-AES, Agilent Technologies
Tubing Peristaltic Pump Tubing
Simple ordering of MP-AES instrument saves time when provisioning your routine instrument maintenance.
  • Convenient kits for easy replacement of peristaltic pump tubing
  • Specific kits for either organic or aqueous samples ensure compatibility with any matrix
  • Simple ordering of peristaltic pump tubing for your MP-AES instrument saves time when planning your instrument maintenance
Simple ordering of peristaltic pump tubing for your MP-AES instrument saves time when provisioning your routine instrument maintenance. Agilent top-quality peristaltic pump tubing allows you to find the appropriate product to handle any matrix. Whether it is PVC, Solvaflex, FPM Viton, or Marprene, Agilent has the peristaltic pump tubing you need for your application.
Convenient kits include replacement peristaltic pump tubing. Specific kits ensure that for either organic or aqueous samples are accommodated for highest performance. Simple ordering of supplies makes routine maintenance easy.