Autosampler Tubing, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
3710052000 5043-0065 3710037100
CA3710052000PK 131.61 CAD
CA3710052000 CAAG5043-0065 CAAG3710037100
Autosampler Tubing, Agilent Technologies
Tubing Peristaltic Pump Tubing
Replacement tubing for autosamplers used with Agilent AAS, ICP-OES, MP-AES and ICP-MS instruments.
  • Works well with solvent and acid-resistant sampling components
  • Easy replacement by the user
A number of different autosamplers have been used with Agilent AA, ICP-OES, MP-AES, and ICP-MS instruments, depending on the specific model. This includes the Agilent SPS 4 used with current instruments, and the SPS 3 and ASX 500 series used with earlier model instruments.