CrossLab Dimpled Liners for GC, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
5190-2296 5190-4006 5190-2297
CA5190-2296EA 110.9 CAD
CA5190-2296 CA5190-4006 CA5190-2297
CrossLab Dimpled Liners for GC, Agilent Technologies
Chromatography Supplies Chromatography Inlet Liners
We designed our 2 mm dimpled liner to be used in the Agilent MultiMode Inlet (MMI) when analyzing heavy matrix samples.
  • Cost-effective for heavy matrix samples
  • Large inert surface area spreads sample, giving longer usable life
  • Exclusive transparent deactivation
  • Recommended for heavy matrix samples, pesticides in food extracts and environmental extracts
We designed our 2 mm dimpled liner to be used in the Agilent MultiMode Inlet (MMI) when analyzing heavy matrix samples such as food or environmental extracts. The liner has been optimized for coldMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable