CrossLab Needle Wash Supplies for HPLC, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
5065-4445 G5668-87200
CA76514-672EA 82.4 CAD
CA76514-672 CAG5668-87200
CrossLab Needle Wash Supplies for HPLC, Agilent Technologies
Syringe Needles
Supplies for needle wash on Agilent autosamplers.
  • Using non-Agilent supplies can considerably shorten lifetime and increases potential baseline noise
The Agilent autosampler is designed to deliver accurate measurements, precise injection volumes, low carryover, and high-quality data. Needle wash is often required to ensure minimum carryover in order to avoid ghost peaks, inaccurate quantitation, or other chromatographic issues. High-quality supplies help maintain the optimal performance of the needle wash process.