Capillary Tubing and Nebulizer Maintenance Supplies, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD) CA
9910024800 9910044000
CA9910024800EA 70.82 CAD
CA9910024800 CA9910044000
Capillary Tubing and Nebulizer Maintenance Supplies, Agilent Technologies
Tubing Chromatography Tubing
Narrow bore and wide bore capillary tubing for various applications, solvents, and sample types.
  • Choose the type of capillary tubing to optimize the performance required for your application with your Agilent nebulizer
  • Use narrow bore capillary tubing (0.38 mm Int.Ø) for aqueous solutions and samples that have relatively low levels of dissolved solids
  • Use wide bore capillary tubing (0.64 mm Int.Ø) for samples that have high levels of dissolved salts and organic solvents
The performance of Agilent nebulizers can be adjusted by switching the type of capillary tubing fitted to the nebulizer.;Precut lengths of narrow bore capillary tubing (0.38 mm Int.Ø) with a flared end is recommended for use with barrel nebulizers in Hi-vac mode and with aqueous solutions. This tubing is also recommended for samples that have relatively low levels of dissolved solids
Wide bore capillary tubing (0.64 mm Int.Ø) is recommended for use with barrel nebulizers in the high solids position and with samples that have high levels of dissolved salts. This tubing is also recommended for use with the adjustable nebulizer when aspirating organic solvents.