Replacement Shelf and Polyethylene Trays/Sumps for Justrite® Safety Cabinets, Justrite®
Supplier: Justrite
29050 29051 29052 29053 29054 29055 29057 29058 29059 29060 29061 29062 29063
76499-344EA 214.23 CAD
76499-344 76499-346 76499-348 76499-350 76499-352 76499-354 76499-356 76499-358 76499-360 76499-362 76499-364 76499-366 76499-368
Replacement Shelf and Polyethylene Trays/Sumps for Justrite® Safety Cabinets, Justrite®
Furniture Shelves
Polyethylene trays over shelves add corrosion protection when storing corrosive liquids.
  • Resistant to aggressive chemicals
  • Polyethylene trays sit on top of galvanized steel shelves
  • Slightly curved edge provides additional protection
  • Patented SpillSlope® galvanized steel shelves direct spills to back and bottom of a leakproof sump
Justrite galvanized steel shelves hold up to 159 kg (350 lbs.) and adjust on 76 mm (3") centers to meet specific workflow needs. Customize your cabinet by adding shelves to increase storage space and accommodate different sized containers. Place polyethylene trays over shelves for corrosion protection when storing corrosive liquids.