MicroCNX™ Connectors, CPC

Supplier: CPC (Colder Products Company)
CNX17002HT CNX17003HT CNX17001HT
76516-390EA 52.54 CAD
76516-390 76516-386 76516-388
MicroCNX™ Connectors, CPC
Fittings and Connectors
Aseptic micro-connectors that provide a simple, efficient method of connecting tubing for small-format biomanufacturing assemblies. MicroCNX™ connectors are the modern alternative to the cumbersome, industrial process of tube welding.

  • PINCH-CLICK-PULL: Intuitive three-step connection process reduces risk of operator error
  • Easy to use: Lowers risk of operator error and related performance, reliability and safety concerns
  • Genderless: Eases single-use systems specifications with one part number for both halves
  • CPC Click: Audible confirmation of assembly with no additional hardware require

The MicroCNX™ line is engineered specifically for the challenging conditions of biologic media transfer in bioprocessing, cell therapy and gene therapy applications.

Autoclave: One cycle up to 130 °C (266 °F) for 60 minutes
Sterilization: Up to 50 kGy gamma-irradiation
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR