Agarose, highly purified

Supplier: Cytiva

17-0554-02 17-0554-02
CA95040-294LEA 872.78 CAD
CA95040-294L 95040-294
Agarose, highly purified
Agarose

Agarose NA has been prepared to give the optimal combination of these properties: Reliable separation of nucleic acids; low inhibition of enzyme activity on recovered material; and high gel strength.


  • Agarose with low electroendosmosis (EEO) is recommended for most DNA/RNA applications.
  • Suitable for all analytical separations as well as most preparative methods.


Agarose with low electroendosmosis (EEO) is recommended for most DNA/RNA applications. If separated nucleic acids are to be used as substrates for enzymes (e.g., ligase and restriction endonucleases), a special highly purified agarose such as Agarose NA should be used.
For preparative work, an agarose with low gelling temperature should be used. Agarose Prep has low melting and gelling points.

Melting Pt: 60…90 °C
Storage Temperature: Ambient
MDL Number: MFCD00081294
CAS Number: 9012-36-6
EINECS: 232-731-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR