Corepoint Scientific® White Diamond Series Refrigerator and Freezer Combination Units, Horizon Scientific
Supplier: Horizon Scientific
PRF102WWW/030 PRF102WWW/0 PRF102WWG/040 PRF102WWG/030 PRF102WWG/0
76516-560EA 7003.84 CAD
76516-560 76516-562 76516-564 76516-566 76516-568
Corepoint Scientific® White Diamond Series Refrigerator and Freezer Combination Units, Horizon Scientific
Refrigerators
These efficient refrigerator and freezer combo units provide quality cold storage in laboratory settings.
  • Microprocessor temperature controller
  • Digital temperature display
  • Audible and visual high and low temperature alarms
  • Remote alarm contacts
  • Keyed door lock
  • Probe access port
  • Natural, hydrocarbon refrigerant
  • White cabinet and door
These White Diamond combo units offer a cold-storage solution for a variety of laboratory needs. Units feature a series of temperature alarms to keep stored products at peak effectiveness. All models are equipped withMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable Safer Human and Environmental Health,Energy Efficient Safer Human and Environmental Health,Energy Efficient Safer Human and Environmental Health,Energy Efficient Safer Human and Environmental Health,Energy Efficient Safer Human and Environmental Health,Energy Efficient
Cat. No. 76516-560 76516-562 76516-564 76516-566 76516-568
Defrost Manual defrost (freezer); cycle defrost (refrigerator) Manual defrost (freezer); cycle defrost (refrigerator) Manual defrost (freezer); cycle defrost (refrigerator) Manual defrost (freezer); cycle defrost (refrigerator) Manual defrost (freezer); cycle defrost (refrigerator)
Shelves/drawers 3 adjustable shelves (freezer); 3 adjustable shelves (refrigerator) 2 fixed shelves (freezer); 3 adjustable shelves (refrigerator) 3 adjustable shelves (freezer); 3 adjustable shelves (refrigerator) 3 adjustable shelves (freezer); 3 adjustable shelves (refrigerator) 2 fixed shelves (freezer); 3 adjustable shelves (refrigerator)
Door type 1 solid swing door (refrigerator); 1 solid swing door (freezer) 1 solid swing door (refrigerator); 1 solid swing door (freezer) 1 glass swing door (refrigerator); 1 solid swing door (freezer) 1 glass swing door (refrigerator); 1 solid swing door (freezer) 1 glass swing door (refrigerator); 1 solid swing door (freezer)
Electrical 115 V, 60 Hz; 2.2 A, ¼ HP (freezer); 1.3 A, ⅙ HP (refrigerator) 115 V, 60 Hz; 1.4 A, ⅛ HP (freezer); 1.6 A, ⅙ HP (refrigerator) 115 V, 60 Hz; 2.2 A, ¼ HP (freezer); 1.3 A, ⅙ HP (refrigerator) 115 V, 60 Hz; 2.2 A, ¼ HP (freezer); 1.3 A, ⅙ HP (refrigerator) 115 V, 60 Hz; 1.4 A, ⅛ HP (freezer); 1.3 A, ⅙ HP (refrigerator)
Weight 85 kg (188 lbs.) 84 kg (186 lbs.) 85 kg (188 lbs.) 85 kg (188 lbs.) 84 kg (186 lbs.)