SpeedVac™ Refrigerated Centrifugal Vacuum Concentrator, SRF110, Thermo Scientific
Supplier: Thermo Fisher Scientific
SRF110-115 SRF110-230
76499-958EA 21646 CAD
76499-958 76499-960
SpeedVac™ Refrigerated Centrifugal Vacuum Concentrator, SRF110, Thermo Scientific
Concentrators
SpeedVac™ SRF110 modular refrigerated vacuum concentrator provides flexibility any benchtop space.
  • Ideal for evaporation of non-aggressive, aqueous based solvents, buffers, HPLC fractions, etc.
  • Three preset programs with option to store up to nine programs
  • Made of powder coated steel with aluminum coated chamber
  • Temperature range −4 to +100 °C (1 °C increment)
  • Self-aligning and quick release rotor mechanism
  • F Gas compliance: Yes (EPA-SNAP and EU)
The unique programming capabilities to maintain chamber temperature pre and post sample run to preventMore Product Information