VWR® Performance Series Glass Door Laboratory Refrigerators
Supplier: VWR International
76514-220 76514-222 76514-224 76514-226 76514-228 76514-230 76514-232 76514-234 76514-236 76514-238
76514-220EA 13631.85 CAD
76514-220 76514-222 76514-224 76514-226 76514-228 76514-230 76514-232 76514-234 76514-236 76514-238
VWR® Performance Series Glass Door Laboratory Refrigerators
Refrigerators
These performance glass door refrigerators offer top-quality features in laboratory cold storage with advanced data logging and superior temperature control.
  • Touch screen digital temperature display and alarm module
  • Extensive alarms including audible, visual, senor error, power failure, low battery, and door ajar alarms
  • Temperature data logging and USB port for data transfer and software updates
  • Microprocessor temperature controller for precise temperature regulation
  • Keyed door lock, LED interior lighting, and adjustable shelving
  • Natural, hydrocarbon refrigerant
  • ENERGY STAR certified*
These VWR® high-performance laboratory refrigerators are designed with extensive unit alarms, touch-screen displays, and advanced capabilities for temperature data logging and transfer. All units utilize natural, hydrocarbon refrigerant for environmental health and economic efficiency.
All units include a three year parts and labor warranty, plus an additional two year compressor parts warranty.
Certifications: ETL listed.
*Select models ENERGY STAR certified: 76514-222, 76514-224, 76514-228, 76514-234