VWR® Standard Series Glass Door Laboratory Refrigerators
Supplier: VWR International
76514-260 76514-262 76514-264 76514-266 76514-268 76514-270 76514-272 76514-274 76514-276 76514-278
76514-260EA 8958.82 CAD
76514-260 76514-262 76514-264 76514-266 76514-268 76514-270 76514-272 76514-274 76514-276 76514-278
VWR® Standard Series Glass Door Laboratory Refrigerators
Refrigerators
These standard glass door laboratory refrigerators offer cold storage solutions at an economical pricepoint.
  • Microprocessor temperature controller
  • Digital temperature display
  • Audible and visual temperature alarms
  • Probe access port
  • Keyed door lock
  • Adjustable shelving
  • Natural, hydrocarbon refrigerant
  • ENERGY STAR certified*
These VWR® standard series refrigerators utilize microprocessor temperature controllers, digital temperature displays, and temperature alarms. The are designed for multi-functional use in laboratory settings. All models also come built with probe access, adjustable shelving, and interior lighting. Units utilize natural, hydrocarbon refrigerant for environmental health and economic efficiency.
All units include a two year parts and labor warranty, plus an additional three year compressor parts warranty.
Certifications: ETL listed.
*Select models ENERGY STAR certified: 76514-262, 76514-264, 76514-266, 76514-274