VWR® Plus Laboratory Freezers, Stainless Steel
Supplier: VWR International
76514-214 76514-216 76514-218
76514-214EA 10762.79 CAD
76514-214 76514-216 76514-218
VWR® Plus Laboratory Freezers, Stainless Steel
Freezers Laboratory / General Purpose Freezers
These VWR® Plus Series stainless steel freezers feature a multi-functional design for laboratory use.
  • Microprocessor temperature controller
  • Digital temperature display
  • Audible and visual temperature alarms
  • Probe access port
  • Keyed door lock
  • Forced draft air circulation
  • Natural, hydrocarbon refrigerant
These VWR® freezers utilize state-of-the-art technology including microprocessor temperature controllers, digital temperature displays, and a series of temperature alarms. All models are equipped with probe access ports, adjustable shelving, and interior lighting. Units utilize natural, hydrocarbon refrigerant for environmental health and economic efficiency.
All units include a two year parts and labor warranty, plus an additional three year compressor parts warranty.
Certifications: ETL listed.