VWR® Plus Series Glass Door Chromatography Refrigerators
Supplier: VWR International
76514-298 76514-300 76514-302 76514-304 76514-306 76514-308 76514-310 76514-312 76514-314
76514-298EA 13100.3 CAD
76514-298 76514-300 76514-302 76514-304 76514-306 76514-308 76514-310 76514-312 76514-314
VWR® Plus Series Glass Door Chromatography Refrigerators
Refrigerators
These VWR® Plus Series glass door chromatography refrigerators offer laboratory cold storage with superior temperature control.
  • Microprocessor temperature controller
  • Digital temperature display and alarm module
  • Audible, visual, senor error, and power failure alarms
  • One GFCI protected duplex electrical outlet and two 2" access ports with self-closing covers
  • Keyed door lock, LED interior lighting, and adjustable shelving
  • Alarm validation and alarm mute with ring back
  • Natural, hydrocarbon refrigerant
  • ENERGY STAR certified*
These VWR® laboratoryMore Product Information