VWR® Standard Series Glass Door Chromatography Refrigerators
Supplier: VWR International
76514-316 76514-318 76514-320 76514-322 76514-324 76514-326 76514-328 76514-330 76514-332
76514-316EA 10149.83 CAD
76514-316 76514-318 76514-320 76514-322 76514-324 76514-326 76514-328 76514-330 76514-332
VWR® Standard Series Glass Door Chromatography Refrigerators
Refrigerators
These standard glass door chromatography refrigerators offer laboratory cold storage at an economical pricepoint.
  • Microprocessor temperature controller
  • Digital temperature display
  • Audible and visual temperature alarms
  • One GFCI protected duplex electrical outlet and one 2" access port with self-closing cover
  • Probe access port
  • Keyed door lock
  • Natural, hydrocarbon refrigerant
  • ENERGY STAR certified*
These VWR® chromatography refrigerators utilize microprocessor temperature controllers, digital temperature displays, and a series of temperature alarms. EachMore Product Information