VWR® Plus Laboratory Refrigerators, Stainless Steel
Supplier: VWR International
76514-202 76514-204 76514-206 76514-208 76514-210 76514-212
76514-202EA 8861.13 CAD
76514-202 76514-204 76514-206 76514-208 76514-210 76514-212
VWR® Plus Laboratory Refrigerators, Stainless Steel
Refrigerators
These VWR® Plus Series stainless steel refrigerators feature a multi-functional design for laboratory use.
  • Microprocessor temperature controller
  • Digital temperature display
  • Audible and visual temperature alarms
  • Probe access port
  • Keyed door lock
  • Forced draft air circulation
  • Natural, hydrocarbon refrigerant
These VWR® refrigerators utilize state-of-the-art technology including microprocessor temperature controllers, digital temperature displays, and a series of temperature alarms. All models are equipped with probe access ports, adjustable shelving,More Product Information
Order Now
SPECIFICATIONS
Cat. No. 76514-202 76514-204 76514-206 76514-208 76514-210 76514-212
Environmentally Preferable Safer Human and Environmental Health,Energy Efficient Safer Human and Environmental Health,Energy Efficient Safer Human and Environmental Health,Energy Efficient Safer Human and Environmental Health,Energy Efficient Safer Human and Environmental Health,Energy Efficient Safer Human and Environmental Health,Energy Efficient
Defrost Cycle defrost Cycle defrost Cycle defrost Cycle defrost Cycle defrost Cycle defrost
Shelves/drawers 3 adjustable shelves 6 adjustable shelves 9 adjustable shelves 4 adjustable shelves 8 adjustable shelves 12 adjustable shelves
Door type 1 stainless steel swing door 2 stainless steel swing doors 3 stainless steel swing doors 1 glass swing door 2 swing glass doors 3 swing glass doors
Material Stainless steel Stainless steel Stainless steel Stainless steel Stainless steel Stainless steel
Electrical 115 V, 60 Hz, 3 A, ⅕ HP 115 V, 60 Hz, 4.5 A, ¼ HP 115 V, 60 Hz, 7.5 A, ½ HP 115 V, 60 Hz, 3 A, ⅕ HP 115 V, 60 Hz, 4.5 A, ¼ HP 115 V, 60 Hz, 7.5 A, ½ HP
Weight 129 kg (285 lbs.) 197 kg (435 lbs.) 274 kg (605 lbs.) 147 kg (325 lbs.) 234 kg (515 lbs.) 329 kg (725 lbs.)