LunaScript RT Master Mix Kit (Primer-free), New England Biolabs
Supplier: New England Biolabs (NEB)
E3025S E3025L
CA76509-480EA 196 CAD
CA76509-480 CA76509-482
LunaScript RT Master Mix Kit (Primer-free), New England Biolabs
Nucleic Acid Reagents RT-PCR and cDNA Reagents
LunaScript RT Master Mix Kit (Primer-free) is an optimized master mix containing all the necessary components for first strand cDNA synthesis except for primers. The mix is compatible with random primers, oligo dT primers, and gene-specific primers enabling maximum cDNA synthesis flexibility.
  • Single-tube 5X master mix contains Luna Reverse Transcriptase, dNTPs, and Murine RNase Inhibitor
  • Primer-free mix enables user flexibility for optimal cDNA synthesis
  • Complete first strand cDNA synthesis protocol in less than 15 minutes
  • Non-interfering, visible tracking dye helps to eliminate pipetting errorsIncludes No-RT Control Mix for increased confidence
LunaScript RT Master Mix (Primer-free) features the thermostable Luna® Reverse Transcriptase which supports cDNA synthesis at elevated temperatures. Murine RNase Inhibitor is included to protect template RNA from degradation. In addition, the presence of a blue tracking dye provides a visual indicator for the RT step as well as downstream applications.