WarmStart Multi-Purpose LAMP/RT-LAMP 2X Master Mix (with UDG), New England Biolabs
Supplier: New England Biolabs (NEB)
M1708S M1708L
CA76509-462EA 404 CAD
CA76509-462 CA76509-464
WarmStart Multi-Purpose LAMP/RT-LAMP 2X Master Mix (with UDG), New England Biolabs
Nucleic Acid Reagents End-point PCR Enzymes and Kits
The WarmStart Multi-Purpose LAMP/RT-LAMP 2X Master Mix (with UDG) is designed to provide a simple, one-step solution for Loop-Mediated Isothermal Amplification (LAMP) of DNA or RNA (RT-LAMP) targets.
  • One-step solution for Loop-Mediated Isothermal Amplification (LAMP) of DNA or RNA (RT-LAMP) targets
  • Reduce risk of carryover contamination with thermolabile UDG and dUTP included in the mix
  • Compatible with multiple detection methods
  • WarmStart technology inhibits activity at room temperature
LAMP and RT-LAMP are commonly used isothermal amplification techniques that provide rapid detection of a target nucleic acid using LAMP-specific primers (supplied by the user) and a strand-displacing DNA polymerase. This mix contains a blend of Bst 2.0 WarmStart DNA Polymerase and WarmStart RTx Reverse Transcriptase in an optimized buffer solution. The inclusion of thermolabile UDG prevents carryover contamination, where unintended product of a previous amplification can serve as the substrate of a subsequent reaction.
The WarmStart Multi-Purpose LAMP/RT-LAMP 2X Master Mix (with UDG) is compatible with multiple detection methods, including turbidity detection, real-time fluorescence detection, and end-point visualization.