Borosil® Culture/Petri Dishes, with Cover, Borosilicate Glass 3.3, Foxx Life Sciences

Supplier: Foxx Life Sciences
3160065 3160077 3160081 3160087
76490-718CS 1437.62 CAD
76490-718 76490-720 76490-722 76490-724 76490-726
Borosil® Culture/Petri Dishes, with Cover, Borosilicate Glass 3.3, Foxx Life Sciences
Culture Dishes Petri Dishes
Borosil® Petri dishes have consistent wall thickness for optical performance.

  • Material: Borosilicate glass 3.3
  • Color: Clear
  • Class/Quality grade: Type I

Borosil® brand Dishes are designed to fulfill wide-ranging needs from crystalizing, evaporating, drying, and storing to Petri dishes for culture, assay and micro-biological works. These are ideal for use in routine work. These Dishes are clear and withstand repeat sterilization. Each is formed to ensure even wall thickness and consistent optical performance. Borosil® brand Dishes are safe, durable and economical.

Certifications: Comply with IS 2626.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR