Borosil® Desiccators Vacuum, Stopcock with PTFE Spindle and Porcelain Plate, Borosilicate Glass 3.3, Foxx Life Sciences
Supplier: Foxx Life Sciences
3083041 3083042 3083043 3083044 3083045
76490-704EA 919.44 CAD
76490-704 76490-706 76490-708 76490-710 76490-712
Borosil® Desiccators Vacuum, Stopcock with PTFE Spindle and Porcelain Plate, Borosilicate Glass 3.3, Foxx Life Sciences
Desiccators Desiccators, Glass
Borosil® vacuum desiccators are generally used for the drying of moist products, and for the storage of moisture-sensitive substances to ensure sample stability.
  • Plates are positioned on an internal ledge within the base
  • Specifically for use under a vacuum
  • Comes with a glass stopcock at the top
The base and cover are made of borosilicate glass 3.3 and all Borosil® brand plain desiccators of the same size have interchangeable bodies and covers.
Certifications: Complies with IS 6128.
Order Now
SPECIFICATIONS
Environmentally Preferable