Borosil® Complete Soxhlet Extraction Apparatus with Allihn Condenser and Flask, Foxx Life Sciences

Supplier: Foxx Life Sciences
3840013 3840016 3840020 3840024 3840029 3840030
76490-728EA 338.19 CAD
76490-728 76490-730 76490-732 76490-734 76490-736 76490-738
Borosil® Complete Soxhlet Extraction Apparatus with Allihn Condenser and Flask, Foxx Life Sciences
Extractors Soxhlet and Soxhlet-Type Extractors
Borosil® Soxhlet extraction apparatus with Allihn condenser is used for the separation of an extraction from a liquid or solid state and is ideal for laboratory scale refluxing.

  • Material: Borosilicate Glass 3.3
  • Color: clear
  • Class/Quality grade: Type

This method enables effective separation with relatively small quantities of solvent. There is a higher cooling capacity because of the greater surface area when using the Allihn Condenser. The component parts of the apparatus are provided with interchangeable joints for great flexibility.

Certifications: Complies with IS 10640.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR