VWR® PCR Plates, 96-Well
Supplier: VWR
VWR®
3400-01-521AV 3400-02-521AV 3400-09-521AV 3450-00-521AV 3400-40-521AV 3400C00-521AV 3410-00-521AV 3450-09-521AV 3420-09-521AV 3450T00-521AV 3400-00-521AV 3420T00-521AV 3400-05-521AV 3400-06-521AV 3420-00-521AV
89049-178CS 603.18 CAD
89049-178 83009-668 89218-290 82006-644 76402-846 82006-646 76402-848 37001-272 82006-640 83007-372 83007-374 82006-648 82006-636 82006-638 76446-884
VWR® PCR Plates, 96-Well
Microplates PCR Plates
VWR® 96-Well PCR Plates are compatible with most leading PCR and real-time PCR instruments.
  • Ultra-thin walls to optimize thermal transfer resulting in increased reaction efficiencies
  • Ultra-clear wells for easy visualization of small sample volumes
  • Made of virgin polypropylene which is naturally low-binding without the need to incorporate additives
  • Black alphanumerics are easy-to-read and simplifies sample identification.
  • All plates are certified free of RNAse, DNAse, and human DNA
Compatible with VWR® 8-strip flat caps (83009-684) and bubble caps (20170-000).
For VWR® Real-Time PCR Plates, search 82006-704.