VWR® Antistatic Ionizer
Supplier: VWR International
76485-872 76485-874
76485-872EA 3182.44 CAD
76485-872 76485-874
VWR® Antistatic Ionizer
Ionizers
The VWR® antistatic ionizer aids in neturalizing electric charges in the air and on samples.
  • Neutralizes electric charges in the environment
  • Useful for measurements with non-conducting materials (plastic, glass)
  • Samples can be 5 to 50 cm away from ionizer
The VWR® antistatic ionizer works by removing overflow of positive and negative ions. An inbalance of negative and positivie ions can cause repeatabilty errors during measurements, the VWR® antistatic ionizer works to neutralize the inbalance to aid measurement accuracy.
Order Now
SPECIFICATIONS
Environmentally Preferable