Dissipative V-Groove Vinyl ESD Floor Mats, Transforming Technologies
Supplier: Transforming Technologies
FM336720GY FM33672GY FM348720GY FM34872GY
76489-686EA 2720.27 CAD
76489-686 76489-688 76489-690 76489-692
Dissipative V-Groove Vinyl ESD Floor Mats, Transforming Technologies
Floor Mats Permanent Floor Mats
Designed to provide a static-safe work area for a wide variety of applications where ESD properties are required.
  • Dissipative ESD floor runner
  • V-groove
  • Gray
V-groove floor runner mats can reduce slips and falls by increasing traction, even when exposed to moisture and chemicals. Transforming Technologies’ VinylStat FM3 V-Groove solid floor runners are made from a homogeneous polymer mix of durable PVC.
Order Now
SPECIFICATIONS
Environmentally Preferable