ESD Anti-Fatigue Mats, ⅜", Transforming Technologies
Supplier: Transforming Technologies
FM22436BN FM224720BN FM224720GY FM23660BN FM23660GY FM236720BN FM236720GY FM248720BN FM248720GY
76489-666EA 181.32 CAD
76489-666 76489-668 76489-670 76489-672 76489-674 76489-676 76489-678 76489-680 76489-682
ESD Anti-Fatigue Mats, ⅜", Transforming Technologies
Floor Mats Traction and Anti-Fatigue Floor Mats
VinylStat FM2 is constructed with a closed-cell expanded PVC that results in a cushioning spongy surface and provides a path-to-ground for electrostatic charges.
  • ESD Anti-fatigue matting
  • Brown or gray
  • ⅜" thick
The mat is available in standalone workstation mats or in 60' rolls. Worker fatigue has been shown to lower productivity and morale which has a large impact on profitability. Damage due to static can have a similar drain on the bottom line - solve both problems with the VinylStat FM2 anti-fatigue matting.