Conductive Rubber V-Groove Floor Mats, Black, Transforming Technologies
Supplier: Transforming Technologies
FM102448 FM103648 FM103X32 FM104872 FM104X32
76489-652EA 313.27 CAD
76489-652 76489-654 76489-656 76489-658 76489-660
Conductive Rubber V-Groove Floor Mats, Black, Transforming Technologies
Floor Mats Permanent Floor Mats
This conductive ESD floor runner has a non-slip, grooved surface that provides excellent traction and allows the mat to be easily cleaned.
  • Conductive ESD floor runner
  • V-groove
  • Gray
  • ⅛" thick
The ribbed matting is also very resilient, allowing it to stand up to heavy foot traffic while still maintaining its ESD protection. The material also offers moderate chemical resistance. The matting is available in 4×32' rolls and an assortment of lengths. The Transforming Technologies GrooveSTAT™ Conductive Rubber Matting provides the electrical properties specified by ESDA standards for floor mats. Most common ESD runners available are made with dissipative vinyl, which does not comply with ESD standards.