Environmental Express® Oakton® Ion100 Portable Ion Meters, Cole-Parmer
Supplier: Antyila Scientific
35613-68 35613-65
76485-972EA 765.02 CAD
76485-972 76485-974
Environmental Express® Oakton® Ion100 Portable Ion Meters, Cole-Parmer
Electrochemistry Meters Ion Meters
The perfect combination of the most advanced electronics technology, sensor technology, and software design.
  • Measured parameter and temperature shown on display at same time
  • Stability, calibration, and low battery indicators
  • Display filter improves measurement, speed, and accuracy
  • IP 57 waterproof and dustproof rated – great for use under harsh environments
The new addition to the Environmental Express Oakton product line features updates to measuring water quality easier and faster. The larger LCD display is 2.75×2.0" for a better viewing angle, hasMore Product Information