extraktLAB Isolate Lab Equipment, United Science
Supplier: United Science
10-1052 80-9613
CA76501-242EA 305413.41 CAD
CA76501-242 CA76501-248
extraktLAB Isolate Lab Equipment, United Science
Services
Equipment required to produce clean CBD isolate at a rate of 1.9 kg/hour. Includes 50 L crystallization reactor, ultra-low temperature reactor, water bath, DrainDroyd starter kit and Octolid, pharma grade cone mill grinder, 20 L rotary evaporator, chiller, vacuum pump, 50 L rotary evaporator, process connections, and tubing.
  • Lab scale kit
  • Proprietary formulation methods and processes for THC removal from CBD distillate using crystallization process
  • Installation and commissioning
With this kit users take control over the process, giving users commandMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable