extraktLAB Delta 8 Lab Equipment Start Up Package, United Science
Supplier: United Science
10-1067 80-9627
CA76501-244EA 254511.18 CAD
CA76501-244 CA76501-250
extraktLAB Delta 8 Lab Equipment Start Up Package, United Science
Services
This 50 L reactor is tailored for Delta 8 production. Equipment features air pumps, heater, solvent reservoirs, manual valves. Helium fluid transfer. Overhead stirrer with air motor manual system, 50 L stirred reactor, air motor, heat exchanger coupling, high temperature ultra-fast oil recirculating heater.
  • Scalable industrial start up package
  • Includes prepackaged consumable kit
  • Low cost start up to produce approx. 500 g of Delta 8 per day from CBD isolate
This process will produce Delta 8 oils at approximately 80 to 85% potency. The oil is then remediated to remove any excess Delta 9, or is diluted to keep the Delta 9 below 0.3%.
Accessories information: The Delta 8 Process and Methods Service/Commissioning provides production methods, training, and processes for Delta 8 production from CBD distillate.