9010 Series ESD Jackets, V-Neck Collar, Knit or Snap Cuff, Transforming Technologies
Supplier: Transforming Technologies
JKV9021BK JKV9021LB JKV9021SPBK JKV9021SPLB JKV9021SPWH JKV9021WH JKV9022BK JKV9022LB JKV9022SPBK JKV9022SPLB JKV9022SPWH JKV9022WH JKV9023BK JKV9023LB JKV9023SPBK JKV9023SPLB JKV9023SPWH JKV9023WH JKV9024BK JKV9024LB JKV9024SPBK JKV9024SPLB JKV9024SPWH JKV9024WH JKV9025BK JKV9025LB JKV9025SPBK JKV9025SPLB JKV9025SPWH JKV9025WH JKV9026BK JKV9026LB JKV9026SPBK JKV9026SPLB JKV9026SPWH JKV9026WH JKV9027BK JKV9027LB JKV9027SPBK JKV9027SPLB JKV9027SPWH JKV9027WH JKV9028BK JKV9028LB JKV9028SPBK JKV9028SPLB JKV9028SPWH JKV9028WH JKV9029BK JKV9029LB JKV9029SPBK JKV9029SPLB JKV9029SPWH JKV9029WH
76489-836EA 91.77 CAD
76489-836 76489-838 76489-840 76489-842 76489-844 76489-846 76489-848 76489-850 76489-852 76489-854 76489-856 76489-858 76489-860 76489-862 76489-864 76489-866 76489-868 76489-870 76489-872 76489-874 76489-876 76489-878 76489-880 76489-882 76489-884 76489-886 76489-888 76489-890 76489-892 76489-894 76489-896 76489-898 76489-900 76489-902 76489-904 76489-906 76489-908 76489-910 76489-912 76489-914 76489-916 76489-918 76489-920 76489-922 76489-924 76489-926 76489-928 76489-930 76489-932 76489-934 76489-936 76489-938 76489-940 76489-942
9010 Series ESD Jackets, V-Neck Collar, Knit or Snap Cuff, Transforming Technologies
Workwear
9010 Series jackets are fabricated from lightweight 90% polyester and 10% carbon fabric for unmatched operator comfort.
  • 90% polyester, 10% carbon
  • Level 3 ESD jacket
  • Maintains consistent continuity after 100 wash cycles
  • V-neck collar
  • Snap-front closure
  • 3 patch pockets
The 9010 Series jackets are fabricated from lightweight 90% polyester and 10% carbon fabric for unmatched operator comfort. Jackets feature a snap-front closure, three roomy patch pockets, and v-neck style collar. The 9010 Series garments maintain consistent continuity readings after 100 wash cycles. Jackets are available with ESD knit cuffs or snap cuffs. Available in blue, black, white, teal, and maroon.